Fire Blanket, Sellstrom®
Catalogue # 33000-159
Fournisseur: Sellstrom
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Fire Blanket
- Cat. no.:33000-159
- Supplier No.:97450
Spécifications
À propos de cet article
Washable, lightweight, woven fiberglass emergency fire blanket is designed to prevent further fire accidents in the lab.
- Sparks, flash fires, and flying particles can be easily extinguished
- Adequately sized to wrap a fire victim
- To clean, wash with soft sponge and air dry only
- Sewn-on grommets allow for hanging during cleaning process
Complete with red herculite pouch for wall hanging. Velcro® pull tab on bottom releases 1.5×1.8 m (5×6') blanket.