Eyewash with Drench Hose, Speakman®
VF-0927-SE
: Speakman
- Description:Eyewash with Drench Hose
- Cat. no.:VF-0927-SE
- Supplier No.:SE-927
The eyewash features a stay-open valve and twin aerated sprays with flip-top dust caps and automatic flow control.
- Eyewash is supplied with 1.8 m (6') of reinforced hose
Designed for countertop mounting, the unit can also be used as a handheld drench hose.
: Meets ANSI Z358.1 standard.