Enclosed Base Worktables, Advance Tabco®
Enclosed base worktables with sliding doors are constructed with 14-gauge, Type 304 stainless steel tops and 18-gauge, Type 304 stainless steel bodies.
- Tables have a 2.5 cm (1") adjustable metal bullet feet
- Standard working height is 90 cm (35½")
Tops have a 4.1 cm (1⁵/₈") sanitary-rolled rim on front edge. Sides and rear edges are flat and turned down on a 90° angle for alignment with walls or other equipment.
Frequently Bought Together