ContactUsLinkComponent
LabExact® Grade TCLP 0.7 µm Binderless Glass Microfiber Filters
Fournisseur:  I.W. TREMONT CO., INC.
New Product
Certificats
undefined
LabExact® Grade TCLP 0.7 µm Binderless Glass Microfiber Filters
Fournisseur:  I.W. TREMONT CO., INC.
New Product
Spécifications
  • Catalog No:
  • 77591-584
  • 77591-592
  • 77591-590
  • 77591-588
  • 77591-586
  • 77591-582
  • Water flow rate:
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • Airflow rate:
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • Retention:
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm