ContactUsLinkComponent
LabExact® Grade A83 0.5 µm Binderless Glass Microfiber Filters
LabExact® Grade A83 0.5 µm Binderless Glass Microfiber Filters
New Product
Certificates
undefined
LabExact® Grade A83 0.5 µm Binderless Glass Microfiber Filters
New Product
Specifications
  • Catalog No:
  • 77590-370
  • 77590-368
  • 77590-366
  • 77590-384
  • 77590-382
  • 77590-380
  • 77590-378
  • 77590-364
  • 77590-376
  • 77590-362
  • 77590-374
  • 77590-360
  • 77590-372
  • 77590-358
  • Water flow rate:
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • 84 s/100 ml, 47 mm Ø @ 10 mbar
  • Airflow rate:
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • 31 s/100 ml/sq. in.
  • Retention:
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm
  • 0.5 µm