Biotrate Digital Burettes
Biotrate Digital Burettes
Supplier:   Sartorius
Biotrate Digital Burettes
Supplier:  Sartorius
Certificates
3 Options Available Below
Product Details & Documents
Biotrate is a premium digital burette equipped with sophisticated functions for easy and convenient titration. Biotrate offers high chemical resistance and it is a smart solution for titrating various liquids accurately and safely.
  • Manual titration of liquids
  • Extremely high chemical resistance
  • Smooth and light to operate with soft touch operating wheels
  • 360° rotating dispensing head
  • Universal connector at the back of the unit for drying tubes and filters
  • User-replaceable valves
  • Long life CR2032 battery
  • Wide range of accessories
Media recirculation system that allows unused media to be recovered from the device and returned to the bottle. Easy to attach aspiration tube with secure threaded connection. Autoclavable liquid pathway (121 ºC, 20 min, 1 bar/100 kPa). Readability 0.01 ml.
Supplied with UV-protected replacement window for light sensitive media protection. Easy custom adjustment for various liquids.
Materials used are Piston: Compound Halar (ECTFE), Cylinder: Borosilicate glass 3.3, Valve block: Halar (ECTFE), Valve mantle: Halar (ECTFE), Valve ball: Borosilicate glass 3.3, Valve seat: Borosilicate glass 3.3, Valve spring: Hastelloy, Tubes: FEP | Sealing rings: PTFE.
Transportation and long-term storage temperature conditions: 0 to 40 ºC.
Limitations: The accuracy and precision values are valid under the controlled conditions described in ISO 8655.
Test temperature: 15 to 30 ºC ±0.5 ºC Relative humidity: >50%.
Certifications: CE certified.
Delivery information: A45 threaded bottle fitting and A32, A38, S40 bottle adapters included. Supplied with 2 years warrenty.
Product Family Options
Informations sur le produit
  • Volume
    Availability
    Price
  • 10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,541.94
    $2,541.94
    Each
       $2,541.94
    Supplier Number: LH-723080
  • 20 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,541.94
    $2,541.94
    Each
       $2,541.94
    Supplier Number: LH-723081
  • 50 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,541.94
    $2,541.94
    Each
       $2,541.94
    Supplier Number: LH-723082
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Dispensing head, adjustable, 0.4 m, FEP
    Dispensing head, adjustable, 0.4 m, FEP
    CA76460-546
    All Biotrate models
    Log in pour la disponibilité
    Each - $110.91
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Dispensing head, standard, FEP
    Dispensing head, standard, FEP
    CA76460-542
    All Biotrate models
    Log in pour la disponibilité
    Each - $62.28
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Dispensing head, fine tip, FEP
    Dispensing head, fine tip, FEP
    CA76460-544
    All Biotrate models
    Log in pour la disponibilité
    Each - $84.07
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Bottle support, silicone, for bottles, 75 - 120 mm
    Bottle support, silicone, for bottles, 75 - 120 mm
    CA76460-610
    All Biotrate models
    Log in pour la disponibilité
    Each - $276.17
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-568
    Prospenser 1, 2, 5 and 10 ml models
    Log in pour la disponibilité
    Each - $25.49
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-572
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $14.31
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-590
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $51.09
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-548
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $162.13
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-576
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $26.94
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-564
    All Prospenser Plus, Biotrate models, Prospenser 30 and 60 ml
    Log in pour la disponibilité
    Each - $25.49
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-584
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $51.09
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-574
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $26.94
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-536
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $62.28
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-550
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $225.84
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-586
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $51.09
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-592
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $51.09
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-570
    Prospenser 1, 2, 5 and 10 ml models
    Log in pour la disponibilité
    Each - $31.75
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-538
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $168.82
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-582
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $26.94
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-594
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $64.85
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-580
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $26.94
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-540
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $168.82
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-588
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $51.09
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-566
    All Prospenser Plus, Biotrate models, Prospenser 30 and 60 ml
    Log in pour la disponibilité
    Each - $31.75
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • CA76460-578
    All Prospenser Plus, Biotrate models
    Log in pour la disponibilité
    Each - $26.94