Individual Precision Analytical Weights, Class 1
Individual Precision Analytical Weights, Class 1
Supplier:   Sartorius
Individual Precision Analytical Weights, Class 1
Supplier:  Sartorius
Certificates
Individual Precision Analytical Weights, Class 1
14 Options Available Below
Product Details & Documents
These Sartorius calibration weights are sold individually with a choice of No Certificate, Traceabililty Certificate, or with a Certificate of Calibration. Your choice of certificates determine the price of your calibration weight.
ASTM: ASTM stands for American Society of Testing and Materials. The ASTM is an international organization that establishes and develops methods for testing materials. ASTM weights and weight sets are made to the tolerance, construction, finish, and magnetism set by them. ASTM requires that weights be made of one piece (no fillers or air to compensate for small adjustments). ASTM weights are recommended for calibrating other weights or high resolution balances.
Traceable Certificate: Many weights and weight sets are available with traceable certificates as an additional option. These certificates are issued by laboratories and manufacturers that are accredited by NIST/NVLAP, ASTM, and/or OIML depending on the type of weight, manufacturer, and class. These certificates are traceable to NIST and contain information such as type of weight, weight class, density, uncertainty, and environmental conditions at the time of calibration.
NVLAP: The National Voluntary Laboratory Accreditation Program (NVLAP). This program evaluates and accredits laboratories in their respective fields and ensures they meet ISO standards.
Product Family Options
Without certification
Informations sur le produit
  • Certification
    Weight
    Availability
    Price
  • Without certification
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $123.82
    $123.82
    Each
       $123.82
    Minimum order quantity is 1
    Supplier Number: WI005MAC1N
  • Without certification
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $182.26
    $182.26
    Each
       $182.26
    Minimum order quantity is 1
    Supplier Number: WI020GAC1N
  • Without certification
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $297.00
    $297.00
    Each
       $297.00
    Minimum order quantity is 1
    Supplier Number: WI200GAC1N
With traceable certification
Informations sur le produit
  • Certification
    Weight
    Availability
    Price
  • Traceable
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $199.93
    $199.93
    Each
       $199.93
    Minimum order quantity is 1
    Supplier Number: WI005MAC1T
  • Traceable
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $259.88
    $259.88
    Each
       $259.88
    Minimum order quantity is 1
    Supplier Number: WI020GAC1T
  • Traceable
    5000 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,261.25
    $2,261.25
    Each
       $2,261.25
    Minimum order quantity is 1
    Supplier Number: WI005KAC1T
With NVLAP certification
Informations sur le produit
  • Certification
    Weight
    Availability
    Price
  • NVLAP
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $244.23
    $244.23
    Each
       $244.23
    Minimum order quantity is 1
    Supplier Number: WI001GAC1C
  • NVLAP
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $281.72
    $281.72
    Each
       $281.72
    Minimum order quantity is 1
    Supplier Number: WI005GAC1C
  • NVLAP
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.50
    $220.50
    Each
       $220.50
    Minimum order quantity is 1
    Supplier Number: WI005MAC1C
  • NVLAP
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.50
    $220.50
    Each
       $220.50
    Minimum order quantity is 1
    Supplier Number: WI010MAC1C
  • NVLAP
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $281.72
    $281.72
    Each
       $281.72
    Minimum order quantity is 1
    Supplier Number: WI020GAC1C
  • NVLAP
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $291.38
    $291.38
    Each
       $291.38
    Minimum order quantity is 1
    Supplier Number: WI030GAC1C
  • NVLAP
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.50
    $220.50
    Each
       $220.50
    Minimum order quantity is 1
    Supplier Number: WI100MAC1C
  • NVLAP
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $546.38
    $546.38
    Each
       $546.38
    Minimum order quantity is 1
    Supplier Number: WI200GAC1C