À propos de cet article
A versatile and reliable tool that will revolutionize your circuit testing experience.
This compact and portable voltmeter is designed to be user-friendly
Easy-to-read digital display, you can effortlessly monitor voltage levels with precision and accuracy
Small size (53×21×15 mm) makes it easy to integrate into any circuitry lesson
Comes equipped with a durable 3 V lithium battery
One of the standout features of this voltmeter is its convenient connectivity options. The two alligator clips directly connected to the meter and attached to the main body allow for hassle-free testing. No more fumbling around with loose wires or complicated setups - simply clip and measure.
With a measurement range of 25 V (with a resolution of 0.01 V), this voltmeter offers versatility for a wide range of applications. From small-scale projects to complex circuits. Automatically switchable display: 0.01 V (0~±5.00 V), 0.1 V. To make your purchase even more convenient, we offer a pack of 10 meters at an unbeatable price.