DC Digital Ammeter with Connectors
Fournisseur: GSC International
New Product
Certificats
À propos de cet article
The 3A, 1mA Digital DC Ammeter with high accuracy is small and light enough to connect with any circuit.
- Two alligator clips are directly connected with the meter’s main body for accurate measurements
- A 3 volt lithium battery is included
The Digital DC Ammeter carries electric current even when powered off, providing quick and highly accurate measurements with one-push start-up. Provided with a zero-point adjustment function that ensures an accurate reading, the Digital DC Ammeter displays measured values digitally with plus or minus sign.
Frequently Bought Together