ContactUsLinkComponent
Accessories for Slide Drying Hotplate, Slimline
Catalogue # 77578-306
Fournisseur:  StatLab
New Product
undefined
Accessories for Slide Drying Hotplate, Slimline
Catalogue # 77578-306
Fournisseur:  StatLab
Supplier Number:  SI-SDH-LID
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Slide drying hotplate lid for use with slide drying hot plate
  • Cat. no.:
    77578-306
Spécifications
À propos de cet article
Optional, easy to use drop-on hot plate lid improves heating efficiency and temperature control.
Ordering information: Lid only, hot plate sold separately.