PathFlow PiSmart Laser Cassettes, Threaded
 :   StatLab
PathFlow PiSmart Laser Cassettes, Threaded
 :  StatLab
PathFlow PiSmart Laser Cassettes, Threaded
Pathflow PiSmart laser cassettes are a two piece design with a rear hinge and front tab closure. Threaded cassettes come in stacks of 40 cassettes, and 50 stacks per case of 2000.
These threaded cassettes can be easily and quickly loaded in the PiSmart Laser Printers. These cassettes are a validated consumable for the PiSmart laser cassette printer.
Product Family Options
  • Désignation
    Color
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Blue
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-BL
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Gray
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-GY
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Green
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-GR
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Lavender
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-LV
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Orange
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-OR
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Pink
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-PK
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Tan
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-TN
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    White
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-WH
  • PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Yellow
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
     : CAS-LPI100BX-YL