ContactUsLinkComponent
BoneClones® Economy Human Child Comparative Set
BoneClones® Economy Human Child Comparative Set
  470356-978
New Product
undefined
BoneClones® Economy Human Child Comparative Set
  470356-978
 :  COMP-400
New Product
  • Description:
    Human child comparative set
  • Cat. no.:
    470356-978
An excellent, economical way to study the human child. Cast from the same molds as the regular Bone Clones® skull and postcranial parts but cranial nerve passages and postcranial foramina undrilled.
The teeth color in the economy versions are the same color (same resin) as the rest of the skull, which is a natural bone color.
 : Contains the 6-year-old human child skull, pelvis, femur and humerus.