Crystal Digital PCR® Assay for TP53 (drop-off Y205-R209/V218-E221)
New Product
Assay-specific Crystal Flex Probes and primers for the detection of TP53 mutations and its positive control. TP53 (drop-off Y205-R209/V218-E221) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify mutations between Y205-R209 or V218-E221 of the TP53 gene using the Ruby Chip. TP53 is pivotal in regulating cell divisison, thus an important tumor suppressor gene.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal Flex Probe detection chemistry for flexible assay configuration
- Same set of Crystal Universal Reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, green
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: TP53 Y205-R209 and V218-E221
: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together