Crystal Digital PCR® Assay for NRAS (Q61K, drop-off Q61)
Fournisseur: STILLA TECHNOLOGIES, INC.
New Product
Certificats
À propos de cet article
Assay-specific Crystal Flex Probes and primers for the detection of NRAS mutations and its positive control. NRAS (Q61K, drop-off Q61) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify mutations in Q61 plus 1 mutation of the NRAS gene. NRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal Flex Probe detection chemistry for flexible assay configuration
- Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Green, blue, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: NRAS Q61/Q61K
Caution: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together