SteriPlast® Detectable Spoons | Avantor
SteriPlast® Detectable Spoons
Supplier:   BURKLE INC
SteriPlast® Detectable Spoons
Supplier:  BURKLE INC
Certificates
SteriPlast® Detectable Spoons
SteriPlast® Detectable Spoons
2 Options Available Below
Product Details & Documents
These detectable SteriPlast® spoons, made of PS, are produced in a class seven cleanroom (10000), individually packaged for disposable use and then sterilised by gamma rays. The detectable spoons are available in two sizes, 2.5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon).
  • Can be detected by color scanners, metal detectors or X-rays
  • Individually packaged and sterilized by gamma rays
  • Production, assembly and packaging according to cleanroom class seven
These spoons are not only colored blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
Certifications: Complies with EU food requirements and FDA regulations.
Product Family Options
Informations sur le produit
  • Longueur
    Capacité
    Availability
    Price
  • 127 mm
    2.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $153.70
    $153.70
    Pack of 100
       $153.70
    Supplier Number: 5378-2011
  • 170 mm
    10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $152.25
    $152.25
    Pack of 100
       $152.25
    Supplier Number: 5378-2012