SteriPlast® Detectable Scoops
Supplier:   BURKLE INC
SteriPlast® Detectable Scoops
Supplier:  BURKLE INC
Certificates
SteriPlast® Detectable Scoops
SteriPlast® Detectable Scoops
8 Options Available Below
Product Details & Documents
These detectable SteriPlast® scoops, made of blue PS, are produced and assembled in a class seven cleanroom (10,000), individually packaged for disposable use and then sterilized by gamma rays. Detectable scoops are ideal for powders, granulates and pastes.
  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilized by gamma rays
  • Production, assembly and packaging according to cleanroom class seven
These scoops are not only colored blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
 : Complies with EU food requirements and FDA regulations.
Product Family Options
Informations sur le produit
  • Capacité
    Longueur
    Color
    Availability
    Price
  • 25 ml
    141 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $68.47
    $68.47
    Pack of 10
       $68.47
    Supplier Number: 5378-2001
  • 50 ml
    160 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $75.64
    $75.64
    Pack of 10
       $75.64
    Supplier Number: 5378-2003
  • 100 ml
    205 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $80.64
    $80.64
    Pack of 10
       $80.64
    Supplier Number: 5378-2005
  • 150 ml
    216 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.73
    $85.73
    Pack of 10
       $85.73
    Supplier Number: 5378-2007
  • 250 ml
    232 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $107.87
    $107.87
    Pack of 10
       $107.87
    Supplier Number: 5378-2013
  • 500 ml
    280 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $196.01
    $196.01
    Pack of 10
       $196.01
    Supplier Number: 5378-2015
  • 1000 ml
    332 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $302.42
    $302.42
    Pack of 10
       $302.42
    Supplier Number: 5378-2019
  • 2000 ml
    381 mm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $375.27
    $375.27
    Pack of 10
       $375.27
    Supplier Number: 5378-2020