VWR® Cornerstone Workbenches
VWR® Cornerstone Workbenches
Supplier:   Treston
VWR® Cornerstone Workbenches
Supplier:  Treston
Certificates
VWR® Cornerstone Workbenches
VWR® Cornerstone Workbenches
6 Options Available Below
Product Details & Documents
The American-made Treston cornerstone industrial workbench combines ergonomic design, heavy-duty steel construction, and unmatched customization options. With a 1500 lbs. load capacity and a wide range of modular accessories, the Cornerstone is perfect for any workplace.
  • Fast, easy assembly
  • Modern design and ESD compatible
  • Four legged steel structure
  • Height adjustment range 29 to 35", in 1" increments
With 10 year warranty.
Product Family Options
Snow white
Informations sur le produit
  • Désignation
    Weighing Capacity
    Hauteur
    W×D
    Availability
    Price
  • Cornerstone frame
    1000 lbs.
    29 - 35"
    96×24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,465.87
    $1,465.87
    Each
       $1,465.87
    Minimum order quantity is 1
    Supplier Number: CSB2496-SW
  • Cornerstone frame
    1000 lbs.
    29 - 35"
    96×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,455.87
    $1,455.87
    Each
       $1,455.87
    Minimum order quantity is 1
    Supplier Number: CSB3096-SW
  • Cornerstone frame
    1000 lbs.
    29 - 35"
    96×36"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,627.68
    $1,627.68
    Each
       $1,627.68
    Minimum order quantity is 1
    Supplier Number: CSB3696-SW
Light neutral
Informations sur le produit
  • Désignation
    Weighing Capacity
    Hauteur
    W×D
    Availability
    Price
  • Cornerstone frame
    1000 lbs.
    29 - 35"
    96×24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,465.87
    $1,465.87
    Each
       $1,465.87
    Minimum order quantity is 1
    Supplier Number: CSB2496-VWR
  • Cornerstone frame
    1000 lbs.
    29 - 35"
    96×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,455.87
    $1,455.87
    Each
       $1,455.87
    Minimum order quantity is 1
    Supplier Number: CSB3096-VWR
  • Cornerstone frame
    1000 lbs.
    29 - 35"
    96×36"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,627.68
    $1,627.68
    Each
       $1,627.68
    Minimum order quantity is 1
    Supplier Number: CSB3696-VWR