Electric Desks TED
Supplier: Treston
New Product
Certificates
About this item
Treston Electric Desk TED comes with quick, silent, and wide height adjustment allowing for a tailored experience for each individual. This ergonomic sit-stand desk perfectly suits light-duty assembly work, packing applications, R&D, and industrial offices.
- Wide height adjustment range: 24.8 to 50" (25.2" range total)
- Motor adjustable
- Quick and silent adjustment: 1" per sec
- Four sizes: Depth 31.5" and Widths 35.4, 43.3, 59.1 or 70.9"
- Electro Static Discharge (ESD) protected model available
- Desk load capacity: 440 lbs, above-desk add-ons: 176 lbs
- Reliable nordic choice, compliant with standards
The desk can be customized with accessories that further enhance ergonomics, free desk space, and provide order. The table withstands high loads and is stable at all heights.
Ordering information: Installation is required. Assembly videos available.
Frequently Bought Together