Pure uTIP™ Pipette Tips for Universal Pipettors
Supplier: Biotix
New Product
Certificates
About this item
Enhance the performance and accuracy of experiments and improve global health with Pure uTIPs.
- Compatible with universal pipettes
Pure utips are made with pure resin and patented technologies to improve accuracy and precision. Flexfit™ lowers necessary insertion and ejection force by ≈70% to make a secure seal, Blade™ lowers the chance of hanging droplet formation, and each tip is free of RNase, DNase, DNA, endotoxins, and nucleic acids. Same great tip and better for the environment.
Pure resin is made of 100% virgin polypropylene. No additives are released into the environment. Pure resin products also comes in 100% recyclable packaging to helps to advance research and sustainability efforts.
Specifications
- Catalog No:
- 77575-892
- 77575-910
- 77575-908
- 77575-890
- 77575-906
- 77575-888
- 77575-886
- 77575-904
- 77575-918
- 77575-916
- 77575-914
- 77575-912
- 77575-894
- 77575-898
- 77575-896
- 77575-922
- 77575-920
- 77575-902
- 77575-900
- 77575-866
- 77575-864
- 77575-874
- 77575-872
- 77575-870
- 77575-868
- 77575-882
- 77575-884
- 77575-880
- 77575-878
- 77575-876
- Environmentally Preferable:
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Environmental certification:
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic