Cornerstone Workbenches
À propos de cet article
The American-made Treston® Cornerstone industrial workbench combines ergonomic design, heavy-duty steel construction, and unmatched customization options. With a 1500 lbs load capacity and a wide range of modular accessories, the Cornerstone is perfect for any workplace.
- Fast, easy assembly
- Modern design and ESD compatible
- Four legged steel structure
- Up to 1500 lbs. load capacity
- Select frames QuickShip eligible
Adjustment range 29 to 35", in 1" increments (leg extenders also available for taller height adjustment).
Color: Light gray.
With 10 year warranty.
Ordering information: Installation is required. Assembly videos available.
4 Options Available Below
Product Details & Documents
The American-made Treston® Cornerstone industrial workbench combines ergonomic design, heavy-duty steel construction, and unmatched customization options. With a 1500 lbs load capacity and a wide range of modular accessories, the Cornerstone is perfect for any workplace.
- Fast, easy assembly
- Modern design and ESD compatible
- Four legged steel structure
- Up to 1500 lbs. load capacity
- Select frames QuickShip eligible
Adjustment range 29 to 35", in 1" increments (leg extenders also available for taller height adjustment).
Color: Light gray.
With 10 year warranty.
Ordering information: Installation is required. Assembly videos available.
Product Family Options
Informations sur le produit
- DésignationW×DW×D×HWeightAvailabilityPrice
- Supplier Number: CSB2496-49
- Supplier Number: CSB3096-49
- Supplier Number: CSB3696-49
- Supplier Number: CSBLE-49