Cellulose Extraction Thimbles, Grade C300
Cellulose Extraction Thimbles, Grade C300
Supplier:   Sartorius
Cellulose Extraction Thimbles, Grade C300
Supplier:  Sartorius
Certificates
Cellulose Extraction Thimbles, Grade C300
Cellulose Extraction Thimbles, Grade C300
26 Options Available Below
Product Details & Documents
These cellulose thimble are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
  • Composed of high-purity cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
They are free of binders and offer a high purity which ensures consistent and high overall flow rates.
Typical wall thickness are 1.3 mm for inner diameters lower than 35 mm and 1.7 mm for inner diameters higher than 35 mm.
Product Family Options
Informations sur le produit
  • Matière
    Désignation
    Longueur
    Inner Diameter
    Épaisseur de paroi
    Availability
    Price
  • Cellulose
    Cellulose extraction thimble, grade C300
    60 mm
    25 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $161.95
    $161.95
    Pack of 25
       $161.95
    Supplier Number: FT-1201-025060
  • Cellulose
    Cellulose extraction thimble, grade C300
    60 mm
    26 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $158.88
    $158.88
    Pack of 25
       $158.88
    Supplier Number: FT-1201-026060
  • Cellulose
    Cellulose extraction thimble, grade C300
    60 mm
    28 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $262.40
    $262.40
    Pack of 25
       $262.40
    Supplier Number: FT-1201-028060
  • Cellulose
    Cellulose extraction thimble, grade C300
    60 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $168.09
    $168.09
    Pack of 25
       $168.09
    Supplier Number: FT-1201-033060
  • Cellulose
    Cellulose extraction thimble, grade C300
    70 mm
    25 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $161.95
    $161.95
    Pack of 25
       $161.95
    Supplier Number: FT-1201-025070
  • Cellulose
    Cellulose extraction thimble, grade C300
    80 mm
    22 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $168.10
    $168.10
    Pack of 25
       $168.10
    Supplier Number: FT-1201-022080
  • Cellulose
    Cellulose extraction thimble, grade C300
    80 mm
    25 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $249.08
    $249.08
    Pack of 25
       $249.08
    Supplier Number: FT-1201-025080
  • Cellulose
    Cellulose extraction thimble, grade C300
    80 mm
    28 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $168.10
    $168.10
    Pack of 25
       $168.10
    Supplier Number: FT-1201-028080
  • Cellulose
    Cellulose extraction thimble, grade C300
    80 mm
    30 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $249.08
    $249.08
    Pack of 25
       $249.08
    Supplier Number: FT-1201-030080
  • Cellulose
    Cellulose extraction thimble, grade C300
    80 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $296.23
    $296.23
    Pack of 25
       $296.23
    Supplier Number: FT-1201-033080
  • Cellulose
    Cellulose extraction thimble, grade C300
    90 mm
    19 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $161.95
    $161.95
    Pack of 25
       $161.95
    Supplier Number: FT-1201-019090
  • Cellulose
    Cellulose extraction thimble, grade C300
    90 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $249.08
    $249.08
    Pack of 25
       $249.08
    Supplier Number: FT-1201-033090
  • Cellulose
    Cellulose extraction thimble, grade C300
    94 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $168.09
    $168.09
    Pack of 25
       $168.09
    Supplier Number: FT-1201-033094
  • Cellulose
    Cellulose extraction thimble, grade C300
    100 mm
    22 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $262.40
    $262.40
    Pack of 25
       $262.40
    Supplier Number: FT-1201-022100
  • Cellulose
    Cellulose extraction thimble, grade C300
    100 mm
    25 mm
    1.4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $296.23
    $296.23
    Pack of 25
       $296.23
    Supplier Number: FT-1201-025100
  • Cellulose
    Cellulose extraction thimble, grade C300
    100 mm
    28 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $249.08
    $249.08
    Pack of 25
       $249.08
    Supplier Number: FT-1201-028100
  • Cellulose
    Cellulose extraction thimble, grade C300
    100 mm
    30 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $169.95
    $169.95
    Pack of 25
       $169.95
    Supplier Number: FT-1201-030100
  • Cellulose
    Cellulose extraction thimble, grade C300
    100 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $262.40
    $262.40
    Pack of 25
       $262.40
    Supplier Number: FT-1201-033100
  • Cellulose
    Cellulose extraction thimble, grade C300
    100 mm
    40 mm
    1.7 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $272.65
    $272.65
    Pack of 25
       $272.65
    Supplier Number: FT-1201-040100
  • Cellulose
    Cellulose extraction thimble, grade C300
    118 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $174.25
    $174.25
    Pack of 25
       $174.25
    Supplier Number: FT-1201-033118
  • Cellulose
    Cellulose extraction thimble, grade C300
    123 mm
    40 mm
    1.7 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $272.65
    $272.65
    Pack of 25
       $272.65
    Supplier Number: FT-1201-040123
  • Cellulose
    Cellulose extraction thimble, grade C300
    123 mm
    43 mm
    1.8 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $272.65
    $272.65
    Pack of 25
       $272.65
    Supplier Number: FT-1201-043123
  • Cellulose
    Cellulose extraction thimble, grade C300
    130 mm
    33 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $174.25
    $174.25
    Pack of 25
       $174.25
    Supplier Number: FT-1201-033130
  • Cellulose
    Cellulose extraction thimble, grade C300
    150 mm
    35 mm
    1.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $457.15
    $457.15
    Pack of 25
       $457.15
    Supplier Number: FT-1201-035150
  • Cellulose
    Cellulose extraction thimble, grade C300
    150 mm
    40 mm
    1.7 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $272.65
    $272.65
    Pack of 25
       $272.65
    Supplier Number: FT-1201-040150
  • Cellulose
    Cellulose extraction thimble, grade C300
    205 mm
    33 mm
    1.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $259.31
    $259.31
    Pack of 25
       $259.31
    Supplier Number: FT-1201-033205