TAQuest™ qPCR Master Mix for TaqMan Probes *Low ROX*
Fournisseur: AAT BIOQUEST INC
Certificats
À propos de cet article
TAQuest™ qPCR Master mix for TaqMan probes is a ready-to-use 2X solution optimized for qPCR and two-step RT-qPCR compatible with TaqMan gene expression assays.
- Simple and robust protocol
- The optimized composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
TAQuest™ qPCR Master mix for TaqMan probes has been designed to be used for duplex reactions using internal positive controls.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimized light exposure.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Application:qPCR probe