TAQuest™ FAST qPCR Master Mix with Helixyte™ Green *High ROX*
Fournisseur: AAT BIOQUEST INC
Certificats
À propos de cet article
TAQuest™ FAST qPCR Master mix with Helixyte™ green is a ready-to-use 2X solution optimized for qPCR and two-step RT-qPCR.
- Simple and robust protocol
- The optimized composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
The Helixyte green intercalating dye allows rapid DNA detection and analysis without using sequence-specific probes.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimized light exposure.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Application:qPCR dye