TAQuest™ FAST qPCR Master Mix with Helixyte™ Green *Low ROX*
Fournisseur: AAT Bioquest
Certificats
À propos de cet article
TAQuest™ FAST qPCR Master mix with Helixyte™ green is a ready-to-use 2X solution optimized for qPCR and two-step RT-qPCR.
- Simple and robust protocol
- The optimized composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
The Helixyte green intercalating dye allows rapid DNA detection and analysis without using sequence-specific probes.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimized light exposure.
Spécifications
- Application:qPCR dye