Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Catalogue # 77461-454
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Size:2
- Cat. no.:77461-454
- Includes:2 labellings
Spécifications
À propos de cet article
Microscale optimized for labelling 25 µg antibody per reaction. APC-iFluor® 750 tandem is an excellent replacement for APC-Cy7 and APC-Alexa Fluor® 750 tandem since they have almost identical spectra.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped with blue ice.
Caution: Research use only.