Cleanroom Labels, Core2Write
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Product Family Options
- DésignationDimensionsPackaging
- Core2Write label, non-sterile50,8×25,4 mm10 labels/sheet/500 sheets/caseCase of 5,000$5,480.00: C2WR-2X1-SHEET-01
- Core2Write label, sterile, irradiated50,8×25,4 mm36 labels/sheet/500 sheets/caseCase of 5,000$5,670.00: C2WR-2X1-SHEET-02
- Core2Write label, sterile, irradiated76,2×25,4 mm18 labels/sheet, 500 sheets/caseCase of 5,000$5,480.00: C2WR-3X1-SHEET-02
- Core2Write label, sterile, irradiated101,6×50,8 mm10 labels/sheet, 500 sheets/caseCase of 5,000$5,480.00: C2WR-4X2-SHEET-02
- Core2Write label, sterile, irradiated101,6×76,2 mm6 labels/sheet, 500 sheets/caseCase of 5,000$5,480.00: C2WR-4X3-SHEET-02