Fetal Bovine Serum, Premium, United States Origin (Tetracycline Negative)
Catalogue # CA76418-028
Fournisseur: Corning
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Sterility:Sterile
- Description:Fetal bovine serum with weight 0.2 lbs.
- Size:50 ml
- Origin:US
- Storage temperature:−4...−10 °C
- Cat. no.:CA76418-028
Spécifications
À propos de cet article
Species: Bovine, bos taurus
Type: Fetal
Treatment: Tetracycline negative
Shelf life: 60 months
- Collected at government approved abattoirs in the United States
- True pooled and triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
- Tetracycline tested to a detection level of 0.001 mg/dl using the AOAC 995.09 testing method