Modular Cart Systems
Fournisseur: Micronova
New Product
Certificats
À propos de cet article
Ergonomic modular cart with smooth maneuverability, and durable, contamination-resistant construction. Interchangeable side rails convert cart to double or triple bucket system and bucket hanger bars are adjustable to accommodate various sized rectangular buckets.
- 304 electropolished stainless steel frame for durability and long life
- Modular rails and adjustable bucket bars to accommodate buckets and configurations
- Cast molded handle holders compatible with any size mop handles
- Stainless steel castors and Prevenz™ antimicrobial wheels
- Foot-operated wheel break to support low-touch systems
- Raised bucket height for ease of use
- Hanging bucket design for greater stability
- Autoclavable
The Micronova modular cart systems feature a contamination-resistant construction, including a stainless-steel frame with a single-piece push bar, cast-molded handle holders, and foot operated wheel break with Prevenz™ antimicrobial wheels. The interchangeable side rails convert the cart to double or triple bucket systems to meet cleaning requirements. Use with any Micronova stainless steel wringers. Compatible with plastic or stainless steel, narrow, or shallow buckets.
Certifications: Certificate of Conformance available upon request.
Delivery: Buckets included. Cart assembly required with easy to follow instructions. Interchangeable side bars sold separately.
Frequently Bought Together