XPR Essential Microbalances
Supplier:   Mettler Toledo
XPR Essential Microbalances
Supplier:  Mettler Toledo
Certificates
XPR Essential Microbalances
XPR Essential Microbalances
2 Options Available Below
Product Details & Documents
METTLER TOLEDO XPR Essential microbalances are highly precise instruments with SmartPan Technology, exceptionally low minimum weight capabilities and a 0.001 mg readability ideal for tasks requiring high precision, such as weighing small quantities of high-value materials or samples. Streamline daily operations with user-friendly features including integrated onscreen guidance and semi-automatic wing doors that open at the push of a button for reduced risk of contamination. METTLER TOLEDO XPR Essential microbalances are the successor to the XSR microbalances.
  • Compact front loader design
  • SmartView terminal
  • 7" color touchscreen
  • Antistatic solutions
  • Multiple interfaces
  • Semi-automatic wing doors
  • Easy clean drip tray
  • Data management
  • Valid and traceable results
The XPR Essential microbalances feature a digital results notepad and storage of SOPs in method library.
Product Family Options
Informations sur le produit
  • Modèle
    Weighing Capacity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • XPR3DUE
    3.2/1.2 g
    32 mm
    0.01/0.001 mg
    6 s
    0.0008 mg
    478×195×196 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $36,830.00
    $36,830.00
    Each
       $36,830.00
    Supplier Number: 30900669
  • XPR16DUE
    12/3.2 g
    32 mm
    0.01/0.001 mg
    6 s
    0.0012 mg
    478×195×196 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44,210.00
    $44,210.00
    Each
       $44,210.00
    Supplier Number: 30904616