Vivaflow® Peristaltic Pump
Peristaltic pump, 100 - 120/200 - 240 V
  CA77590-226
 :   Sartorius
 :  VF-APD0001-1
Vivaflow® Peristaltic Pump
  CA77590-226
 :  Sartorius
Vivaflow® Peristaltic Pump
Vivaflow® Peristaltic Pump
Peristaltic pump, 100 - 120/200 - 240 V
$5,640.96
Each
Recommended for processing samples with Vivaflow® tangential flow filtration (TFF) cassettes, these Watson-Marlow pumps are ideal for ultrafiltration and diafiltration applications.
  • Simple and intuitive set-up
  • Precise flow control
  • Avoid contamination
  • Low maintenance
Peristaltic pumps offer the flexibility, performance, and control needed for other filtration and fluid transfer applications. Each pump includes a power cable with region-specific plug, according to the delivery location. A voltage selector switch can be set for use on either 100 to 120 V, 50/60 Hz or 200 to 240 V, 50/60 Hz power supplies.
  • Désignation:
    Peristaltic pump, 100 - 120/200 - 240 V