ContactUsLinkComponent
Carson Optical Magnifier and Quick Release Bug Catching Tool
  470229-200
undefined
Carson Optical Magnifier and Quick Release Bug Catching Tool
  470229-200
 :  HU-10
  • Description:
    5× magnifier and quick-release bug catching tool
  • Size:
    2.2×1.6×10.5"
  • Cat. no.:
    470229-200
One-hand bug catching tool for kids.
  • Thumb operated
  • Crystal-clear acrylic
  • Weight: 0.25 lbs.
This easy to use device will engage students with field biology in a fun, engaging, and hands-on way. Students will see the smallest details with ease. Simply catch a bug, view, and once you’re finished examining, release.