ROX Dye qPCR Calibration Solution *10000X*
Catalog # 77463-014
Supplier: AAT BIOQUEST INC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:ROX Dye qPCR calibration solution *10000X*
- Size:100 μl
- Cat. no.:77463-014
Specifications
About this item
ROX dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems.
- Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.