ContactUsLinkComponent
HIS Lite™ iFluor® 647 Tris NTA-Ni Complex
Catalogue # 77461-398
Fournisseur:  AAT Bioquest
Numéro CAS:  
undefined
HIS Lite™ iFluor® 647 Tris NTA-Ni Complex
Catalogue # 77461-398
Fournisseur:  AAT Bioquest
Supplier Number:  12618
Numéro CAS:  
Spécifications
  • Description:
    HIS Lite™ iFluor® 647 Tris NTA-Ni complex
  • Size:
    100 μg
  • Cat. no.:
    77461-398
Spécifications
À propos de cet article
iFluor 647-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labeled proteins in cells, solution and solid surfaces.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.