ContactUsLinkComponent
HIS Lite™ Cy3 Tris NTA-Ni Complex
HIS Lite™ Cy3 Tris NTA-Ni Complex
Catalog # 77461-402
Supplier:  AAT Bioquest
CAS Number:  
undefined
HIS Lite™ Cy3 Tris NTA-Ni Complex
Catalog # 77461-402
Supplier:  AAT Bioquest
Supplier Number:  12620
CAS Number:  
Specifications
  • Description:
    HIS Lite™ Cy3 Tris NTA-Ni complex
  • Size:
    100 μg
  • Cat. no.:
    77461-402
Specifications
About this item
Cy3-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labeled proteins in cells, solution and solid surfaces.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.