ContactUsLinkComponent
FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimized for Tyramide and Styramide Imaging*
Catalogue # 77462-260
Fournisseur:  AAT BIOQUEST INC
Numéro CAS:  
undefined
FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimized for Tyramide and Styramide Imaging*
Catalogue # 77462-260
Fournisseur:  AAT BIOQUEST INC
Supplier Number:  44890
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    FluoroQuest™ TSA/PSA Antifade mounting medium *Optimized for Tyramide and Styramide imaging*
  • Size:
    5 ml
  • Cat. no.:
    77462-260
Spécifications
À propos de cet article
FluoroQuest™ TSA/PSA Antifade mounting medium is the only mounting medium that is optimized for tyramide (TSA) and styramide (PSA)-based fluorescence imaging applications.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.