Wheat Germ Agglutinin, XFD750 Labeled *XFD750 Same Structure to Alexa Fluor® 750*
Catalogue # 77462-024
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Description:Wheat germ agglutinin, XFD750 labeled *XFD750 same structure to Alexa Fluor® 750*
- Size:1 mg
- Cat. no.:77462-024
Spécifications
À propos de cet article
XFD750, manufactured by AAT Bioquest, has an identical chemical structure to Alexa Fluor® 750, a trademark of ThermoFisher.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.