ReadiView™ Blue Real-Time PCR Visualization Dye *200X*
77461-618
:
- Description:ReadiView™ blue real-time PCR visualization dye *200X*
- Size:500 tests
- Cat. no.:77461-618
ReadiView™ Blue Real-Time PCR Visualization Dye is a 200X concentrated, ready-to-use, inert dye that enhances the visibility of real-time PCR reactions for more accurate pipetting and plate loading.
- Stable for 12 months
Quality tested in-house.
: For research use only.