ContactUsLinkComponent
HIS Lite™ Cy5 Tris NTA-Ni Complex
  77461-400
 :  
undefined
HIS Lite™ Cy5 Tris NTA-Ni Complex
  77461-400
 :  12619
 :  
  • Description:
    HIS Lite™ Cy5 tris NTA-Ni complex
  • Size:
    100 µg
  • Cat. no.:
    77461-400
Cy5-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labeled proteins in cells, solution and solid surfaces.
  • Stable for 12 months
Quality tested in-house.
 : For research use only.