ContactUsLinkComponent
FAM Dye qPCR Calibration Solution *10000X*
Catalogue # 77463-010
Fournisseur:  AAT BIOQUEST INC
Numéro CAS:  
undefined
FAM Dye qPCR Calibration Solution *10000X*
Catalogue # 77463-010
Fournisseur:  AAT BIOQUEST INC
Supplier Number:  67032
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    FAM dye qPCR calibration solution *10000X*
  • Size:
    100 µl
  • Cat. no.:
    77463-010
Spécifications
À propos de cet article
FAM dye qPCR calibration solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
  • Stable for 12 months
Calibration and verification should be run every six months or as needed.
Caution: For research use only.