ContactUsLinkComponent
Amplite® Tyramide (TSA) Signal Booster
Catalog # 77462-258
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Tyramide (TSA) Signal Booster
Catalog # 77462-258
Supplier:  AAT Bioquest
Supplier Number:  44502
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Amplite® Tyramide (TSA) signal booster
  • Size:
    1000 reactions
  • Cat. no.:
    77462-258
Specifications
About this item
Tyramide signal amplification (TSA) is a technique used to enhance the signal of target molecules, such as antibodies or nucleic acid probes, in immunohistochemistry and in situ hybridization.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.