ContactUsLinkComponent
EO Chex™ Ethylene Oxide Indicator Tape – FDA Cleared
EO Chex™ Ethylene Oxide Indicator Tape – FDA Cleared
Catalog # 77578-300
Supplier:  Propper
New Product
undefined
EO Chex™ Ethylene Oxide Indicator Tape – FDA Cleared
Catalog # 77578-300
Supplier:  Propper
Supplier Number:  26811500
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Dimensions:
    1.9 cm × 50.3 m (³/₄" × 55 yds.)
  • Cat. no.:
    77578-300
Specifications
About this item
Indicator tape for ethylene oxide sterilization.
  • Roll of strong self-adhesive tape printed with Type 1 indicator ink
  • Initial color: Pink; Final color: Orange-brown
  • Free of toxic heavy metals
  • Write on tape or apply pre-printed labels
EO Chex™ Ethylene Oxide (EO/EtO) indicator tape distinguishes between processed and unprocessed packages sterilized in Ethylene Oxide (EO/EtO) gas.
EO Chex™ features indicator ink placed in pink stripes that turn orange-brown after exposure to an ethylene oxide cycle.
EO Chex™ Beige indicator tapes strong adhesive and even coating allow even dispensing and easy application. The specifically formulated universal adhesive provides secure bonding with fabric and non-woven wraps, paper, metal, and glass, yet allows easy removal. Tape will remove cleanly and easily following sterilization.
Packaging: Case of 48 rolls.
Caution: EO Chex™ Ethylene Oxide indicator tape should be stored at or below room temperature (25 °C/77 °F) in its original packaging. Relative Humidity is less than 60%. Keep EO Chex™ away from sunlight and chemicals, cleaning and disinfecting products. The shelf life of this product is 24 months from the date of manufacture, when stored at the recommended conditions.