Single-Stage Lil’pet Micropipettes
Fournisseur: MICROLIT USA
New Product
Certificats
À propos de cet article
Great for controlled gel loading.
- Only the first stop is depressed
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
This fixed-volume pipette features a single stop plunger to expel all liquid from the tip. Only the first stop is depressed, which is the lowest position to draw and dispense the reagent.
Frequently Bought Together