Klinger Brewster's Angle Experiment
Brewster angle apparatus
  470356-826
 :   Klinger
 :  KSCIBAE
Klinger Brewster's Angle Experiment
  470356-826
 :  Klinger
Klinger Brewster's Angle Experiment
Klinger Brewster's Angle Experiment
Brewster angle apparatus
2 741,30 $
Each
Measure Brewster’s angle for a given dielectric dispersive medium.
  • Compact rotating table makes the apparatus extremely smooth to operate
  • Sensitive light sensor (1 lux) enables the measurement of light intensity at various angles of reflection
When un-polarized light is incident on the surface of a dielectric (such as a glass), at a certain angle of incidence the reflected light is completely plane-polarized. This phenomenon was discovered by Sir David Brewster and, thus, the specific angle is called Brewster’s angle or polarization angle. Also, from the experiment, it can be confirmed that the reflected ray and the refracted ray are 90° apart when the incident angle is set at Brewster’s angle.
 : Kit includes optical bench set 0.8 m, light source holder, converging lens holder, adjustable collimating slit holder, rotating table assembly, analyzer square, light sensor holder, power supply for light source, glass block, data processor.
  • Désignation:
    Brewster angle apparatus