Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

XFD647 azide

1 mg
Catalog # 77461-724
Supplier:   AAT Bioquest
 :  1896
XFD647 azide
Catalog # 77461-724
Supplier:  AAT Bioquest
CAS Number:  
XFD647 azide
XFD647 azide

1 mg

CAS Number:  
Price
$344.75
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molecular Weight:1185.29 g/mol
Specifications

Specifications

  • Taille:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.