Cy5 aldehyde
1 mg
  77463-120
 :  9012
Cy5 aldehyde
  77463-120
 :  
Cy5 aldehyde
Cy5 aldehyde
1 mg
 :  
$352.84
Each(1/Itm/Bx)
Cy5 aldehyde is a reactive red fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
  • Stable for 12 months
Quality tested in-house
 : For Research Use Only
  •  :945.04 g/mol
  • Taille:
    1 mg