Welch® Vacuum Pumps
Supplier: Labconco
New Product
Certificates
About this item
Rotary vane vacuum pump.
- Acid-resistant
- Used for samples like acids and solvents
- Applicable products: Freeze dryers
- Value size: ³/₄"
Specifications
- Catalog No:
- 77574-110
- 77574-108
- Ultimate vacuum:
- 2×10ˉ³ mbar
- 2×10ˉ³ mbar
- W×D×H:
- 28.2W×46.2D×23.4 H (11.1W×18.2D×9.2" H)
- 28.2W×46.2D×23.4 H (11.1W×18.2D×9.2" H)
- Electrical:
- 230 V
- 115 V