Eppendorf Xplorer single-channel pipettes provide reliable, ergonomic high-precision liquid handling. With easy operation, five pipetting modes, and advanced charging technology, they minimize errors and boost reproducibility in a range of lab applications.
- Electronic single channel pipettes designed to aspirate and dispense liquid volumes from 0.5 μl to 10 ml
- Intuitive operating concept and ergonomic design
- Multifunctional rocker with ergonomic display angle
- Selection dial and innovative ejector
- Convenient handle and powerful rechargeable battery
- Flexible charging concept
- Spring-loaded tip cone (except 5 and 10 ml)
- Adjustment to external conditions (geographic height, liquids with high density or similar factors)
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
Nine different language settings: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish. Air-cushion pipetting system, with autoclavable lower part and epT.I.P.S.® consumables.
Applications: Pipetting of aqueous liquids.
- Catalog No:
- CA89133-428
- CA89133-426
- CA89133-424
- CA89133-432
- CA89133-430
- CA76420-656
- CA76420-654
- CA89133-434
- CA76420-658
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental certification:
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label