Eppendorf® Xplorer® Plus Move It® Electronic Multi-Channel Pipettes With Adjustable Tip Spacing
Eppendorf® Xplorer® Plus Move It® Electronic multi-channel pipettes with adjustable cones allow easy vessel format changes, cutting up to 50% of your time compared to single-channel pipettes. With several pipetting modes for enhanced productivity and offered in 4, 6, 8 or 12 channels.
- Air-cushion pipettes with variable volume and adjustable tip spacing are available in 4, 6, 8 and 12-channel
- Choose from Mechanical (Research™ plus) or Electronic (Xplorer™ plus) adjustable tip spacing multichannel pipettes
- For Plates (12, 24, 48, 96, and 384-well), Tubes (1.5 and 2 ml), and Agarose gels
- Tip spacing is freely selectable from 4.5 to 33 mm, depending on the pipette variant
- Spacing control for quick switches backwards and forwards between source and destination formats
- Manual adjustment knob for smooth, vibration-free format switches and drip-free liquid transfers
- 360° rotatable pipette head for comfortable readability of display and fast parameter identification
- Tubeless system is safe and reliable and optimally balanced pipette reduces hand fatigue for stress-free operation
Air-cushion pipetting system and autoclavable for user and sample safety (Research™ plus fully, and Xplorer™ plus lower part only).
Applications: Pipetting of aqueous liquids
- Catalog No:
- CA76474-592
- CA76474-596
- CA76474-594
- CA76541-126
- CA76474-600
- CA76541-128
- CA76474-598
- CA76541-130
- CA76541-132
- CA76474-602
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Frequently Bought Together