About this item
Eppendorf Xplorer® multi-channel pipettes with dispensing mode provide a cutting-edge solution for high-throughput liquid handling. Offering the highest levels of simplicity, precision, and reproducibility, these pipettes will help you pipette with more speed, comfort, and ergonomics.
- Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0.5 μl to 10 ml
- Intuitive operating concept and ergonomic design
- Multifunctional rocker and ergonomic display angle
- Selection dial and innovative ejector
- Convenient handle with powerful rechargeable battery
- Flexible charging concept
- Spring-loaded tip cone (except 5 and 10 ml)
- Adjustment to external conditions (geographic height, liquids with high density or similar factors)
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
Air-cushion pipetting system with epT.I.P.S.® consumables and autoclavable lower part. Nine different language settings: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish.
Application: Pipetting of aqueous liquids.
Specifications
- Catalog No:
- CA89133-444
- CA89133-442
- CA89133-448
- CA89133-446
- CA89496-610
- CA89133-436
- CA89133-440
- CA89133-438
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001