Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
CaCl₂ Solution

CaCl₂ Solution

CaCl₂ solution, 25 µl, Enzyme assay, –20 °C
Catalog # 77554-934
Supplier:   Sino Biological
Supplier #:  C02-09-25
CaCl₂ Solution
Catalog # 77554-934
Supplier:  Sino Biological
CAS Number:  
CaCl₂ Solution

CaCl₂ solution, 25 µl, Enzyme assay, –20 °C

CAS Number:  
Price
$27.49
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

25 μl volume of 1 M CaCl₂ solution for different enzyme assays.

Specifications

Specifications

  • Désignation:
    CaCl₂ solution
  • Taille:
    25 µl
  • Température de stockage:
    –20 °C
  • Utilisation:
    Enzyme assay
  • Cat. No.:
    77554-934
  • Storage conditions:
    For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Formulation:
    Frozen solution contains: 1 M CaCl₂ in sterile water.

Specifications